Assay Details
Target Gene Details
Entrez Gene ID: | 7430 |
Gene Name: | ezrin |
Gene Aliases: |
CVIL, CVL, HEL-S-105, VIL2 |
Location: |
Chr.6:158765741-158819424 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 2 - Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| EZR | NM_001111077.1 | NP_001104547.1 | ||
| NM_003379.4 | NP_003370.2 | |||
| AK296738.1 | ||||
| AK299456.1 | ||||
| AK312597.1 | ||||
| AL162086.1 | CAB82418.1 | |||
| BC013903.2 | AAH13903.1 | |||
| BC068458.1 | AAH68458.1 | |||
| DA754543.1 | ||||
| DA783190.1 | ||||
| FJ224318.1 | ||||
| J05021.1 | AAA61278.1 | |||
| X51521.1 | CAA35893.1 |
Target Gene Details
Entrez Gene ID: | 101409257 |
Gene Name: | EZR antisense RNA 1 |
Gene Aliases: |
- |
Location: |
Chr.6:158817979-158822239 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 1 - Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| EZR-AS1 | NR_102425.1 | |||
| AI831881.1 | ||||
| BE552153.1 | ||||
| BX119561.1 |
More Information
Set Membership: |
|

Genomic Map