Assay Details
Target Gene Details
Entrez Gene ID: | 57149 |
Gene Name: | LYR motif containing 1 |
Gene Aliases: |
A211C6.1 |
Location: |
Chr.16:20899626-20925006 on Build GRCh38 |
Assay Gene Location: | Within Exon 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LYRM1 | NM_001128301.2 | 5 | 1306 | NP_001121773.1 |
| NM_001128302.2 | 4 | 1074 | NP_001121774.1 | |
| NM_001302835.1 | 6 | 1250 | NP_001289764.1 | |
| NM_001302836.1 | 6 | 1402 | NP_001289765.1 | |
| NM_020424.4 | 5 | 1225 | NP_065157.1 | |
| NR_126516.1 | 4 | 1147 | ||
| NR_126520.1 | 3 | 929 | ||
| NR_126521.1 | 3 | 915 | ||
| NR_126527.1 | 3 | 743 | ||
| NR_126528.1 | 4 | 1161 | ||
| XM_005255444.1 | 5 | 1127 | XP_005255501.1 | |
| XM_011545900.2 | 5 | 889 | XP_011544202.1 | |
| XM_011545901.2 | 5 | 955 | XP_011544203.1 | |
| XM_011545902.1 | 6 | 1226 | XP_011544204.1 | |
| XM_011545908.2 | 5 | 957 | XP_011544210.1 | |
| XM_017023475.1 | 6 | 1642 | XP_016878964.1 | |
| XM_017023476.1 | 4 | 1540 | XP_016878965.1 | |
| XM_017023477.1 | 4 | 1477 | XP_016878966.1 | |
| XM_017023478.1 | 5 | 947 | XP_016878967.1 | |
| XM_017023479.1 | 5 | 1384 | XP_016878968.1 | |
| XM_017023480.1 | 7 | 1470 | XP_016878969.1 | |
| XM_017023481.1 | 5 | 1099 | XP_016878970.1 | |
| XM_017023482.1 | 4 | 1153 | XP_016878971.1 | |
| AK308690.1 | 3 | 994 | ||
| AL359616.1 | 1 | 327 | ||
| BC017039.1 | 6 | 1250 | AAH17039.1 | |
| BG541469.1 | ||||
| BM546065.1 | 3 | 879 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv457444 | Chr.16:20917903 - 21160552 on Build GRCh38 | Loss |
|
| nsv436835 | Chr.16:18930652 - 21500976 on Build GRCh38 | Insertion |
|

Genomic Map