Assay Details
Target Gene Details
Entrez Gene ID: | 84079 |
Gene Name: | ankyrin repeat domain 27 |
Gene Aliases: |
PP12899, VARP |
Location: |
Chr.19:32597001-32675215 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 26 - Exon 27 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ANKRD27 | NM_032139.2 | NP_115515.2 | ||
| XM_006723412.2 | XP_006723475.1 | |||
| XM_017027349.1 | XP_016882838.1 | |||
| AK054561.1 | ||||
| AL136784.1 | CAB66718.1 | |||
| AL834335.1 | CAD39003.1 | |||
| BC050529.1 | AAH50529.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3644175 | Chr.19:32600959 - 32614757 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map