Assay Details
Target Gene Details
Entrez Gene ID: | 9033 |
Gene Name: | polycystin 2 like 1, transient receptor potential cation channel |
Gene Aliases: |
PCL, PKD2L, PKDL, TRPP3 |
Location: |
Chr.10:100288145-100346472 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 18 - Exon 18 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PKD2L1 | NM_001253837.1 | NP_001240766.1 | ||
| NM_016112.2 | NP_057196.2 | |||
| XM_011540323.2 | XP_011538625.1 | |||
| XM_011540325.2 | XP_011538627.1 | |||
| XM_017016875.1 | XP_016872364.1 | |||
| XM_017016876.1 | XP_016872365.1 | |||
| AF053316.1 | AAD51859.1 | |||
| AF073481.1 | AAD41638.1 | |||
| AF092170.1 | AAD16101.1 | |||
| AF094827.1 | AAD08695.1 | |||
| BC025665.1 | AAH25665.1 | |||
| DQ084244.1 | AAZ31363.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2761622 | Chr.10:100249953 - 100290757 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map