Assay Details
Target Gene Details
Entrez Gene ID: | 27199 |
Gene Name: | oxoglutarate receptor 1 |
Gene Aliases: |
GPR80, GPR99, P2RY15, P2Y15, aKGR |
Location: |
Chr.13:96985719-96995842 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 3 - Intron 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| OXGR1 | NM_080818.3 | NP_543008.3 | ||
| XM_005254043.3 | XP_005254100.1 | |||
| XM_005254044.3 | XP_005254101.1 | |||
| XM_005254045.2 | XP_005254102.1 | |||
| XM_005254046.2 | XP_005254103.1 | |||
| XM_011521077.2 | XP_011519379.1 | |||
| AK122639.1 | ||||
| AK313684.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3633286 | Chr.13:96988346 - 96989756 on Build GRCh38 | Loss |
|
| nsv832693 | Chr.13:96942591 - 97125227 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map