Assay Details
Target Gene Details
Entrez Gene ID: | 4114 |
Gene Name: | MAGE family member B3 |
Gene Aliases: |
CT3.5 |
Location: |
Chr.X:30230436-30237493 on Build GRCh38 |
Assay Gene Location: | Within Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MAGEB3 | NM_002365.4 | 5 | 825 | NP_002356.2 |
| XM_011545513.2 | 4 | 432 | XP_011543815.1 | |
| XM_011545514.2 | 3 | 399 | XP_011543816.1 | |
| AK057441.1 | 5 | 825 | ||
| BC069784.1 | 1 | 147 | AAH69784.1 | |
| BC074756.2 | 1 | 116 | AAH74756.1 | |
| BC126320.1 | 1 | 119 | ||
| BC126322.1 | 1 | 119 |
More Information
Set Membership: |
|

Genomic Map