Assay Details
Target Gene Details
Entrez Gene ID: | 8702 |
Gene Name: | beta-1,4-galactosyltransferase 4 |
Gene Aliases: |
B4Gal-T4, beta4Gal-T4 |
Location: |
Chr.3:119211742-119240946 on Build GRCh38 |
Assay Gene Location: | Within Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| B4GALT4 | NM_003778.3 | 3 | 452 | NP_003769.1 |
| NM_212543.1 | 4 | 535 | NP_997708.1 | |
| XM_005247855.1 | 3 | 510 | XP_005247912.1 | |
| XM_006713798.2 | 4 | 595 | XP_006713861.1 | |
| XM_006713799.2 | 4 | 640 | XP_006713862.1 | |
| XM_006713800.1 | 3 | 487 | XP_006713863.1 | |
| XM_006713801.2 | 5 | 678 | XP_006713864.1 | |
| XM_011513260.1 | 5 | 607 | XP_011511562.1 | |
| XM_017007401.1 | 3 | 425 | XP_016862890.1 | |
| AB024436.1 | BAA75821.1 | |||
| AF020920.1 | 2 | 343 | AAG50147.1 | |
| AF022367.1 | AAC72493.1 | |||
| AF038662.1 | 2 | 343 | AAC39735.1 | |
| AK001006.1 | 4 | 508 | ||
| AK301688.1 | ||||
| AK314425.1 | 3 | 425 | ||
| AY359008.1 | 1 | 151 | AAQ89367.1 | |
| BC004523.2 | 2 | 201 | AAH04523.1 | |
| BC062618.1 | 3 | 674 | AAH62618.1 | |
| BX647468.1 | 3 | 425 | ||
| CR749555.1 | 3 | 435 | CAH18352.1 |
Target Gene Details
Entrez Gene ID: | 100874201 |
Gene Name: | B4GALT4 antisense RNA 1 |
Gene Aliases: |
- |
Location: |
Chr.3:119226486-119235141 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| B4GALT4-AS1 | NR_046574.1 | |||
| DB092185.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv25249 | Chr.3:119136490 - 119281404 on Build GRCh38 | Loss |
|
| esv3597553 | Chr.3:119144924 - 119291446 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map