Assay Details
Target Gene Details
Entrez Gene ID: | 140032 |
Gene Name: | ribosomal protein S4, Y-linked 2 |
Gene Aliases: |
RPS4Y2P |
Location: |
Chr.Y:20756068-20781032 on Build GRCh38 |
Assay Gene Location: | Overlaps - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| RPS4Y2 | NM_001039567.2 | NP_001034656.1 | ||
| AF497481.2 | AAM18074.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv10025 | Chr.Y:18865177 - 20859250 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map