Assay Details
Target Gene Details
Entrez Gene ID: | 6097 |
Gene Name: | RAR related orphan receptor C |
Gene Aliases: |
IMD42, NR1F3, RORG, RZR-GAMMA, RZRG, TOR |
Location: |
Chr.1:151806071-151832450 on Build GRCh38 |
Assay Gene Location: | Within Exon 13 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| RORC | NM_001001523.1 | 10 | 2007 | NP_001001523.1 |
| NM_005060.3 | 11 | 2037 | NP_005051.2 | |
| XM_006711484.3 | 12 | 2820 | XP_006711547.2 | |
| AK097994.1 | 10 | 2021 | ||
| AK128522.1 | 2 | 3999 | ||
| AK223137.1 | 11 | 1963 | BAD96857.1 | |
| AK298460.1 | 12 | 2120 | ||
| AL834219.1 | 10 | 2007 | CAD38900.1 | |
| BC031554.1 | 11 | 1955 | AAH31554.1 | |
| BC110571.2 | 11 | 1989 | AAI10572.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1010398 | Chr.1:151789173 - 151818464 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map