Assay Details
Target Gene Details
Entrez Gene ID: | 151354 |
Gene Name: | family with sequence similarity 84 member A |
Gene Aliases: |
NSE1, PP11517 |
Location: |
Chr.2:14632686-14640046 on Build GRCh38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| FAM84A | NM_145175.2 | 2 | 2610 | NP_660158.2 |
| AJ417080.1 | CAD10038.1 |
Target Gene Details
Entrez Gene ID: | 653602 |
Gene Name: | uncharacterized LOC653602 |
Gene Aliases: |
- |
Location: |
Chr.2:14635091-14650819 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LOC653602 | NM_001291410.1 | NP_001278339.1 | ||
| AK092711.1 | BAC03955.1 |
More Information
Additional Information:
For this assay, SNP(s) [rs79772157] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map