Assay Details
Target Gene Details
Entrez Gene ID: | 55705 |
Gene Name: | importin 9 |
Gene Aliases: |
Imp9 |
Location: |
Chr.1:201829160-201884294 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 1 - Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| IPO9 | NM_018085.4 | NP_060555.2 | ||
| AF410465.1 | AAL01416.1 | |||
| DA861528.1 | ||||
| DB232332.1 |
Target Gene Details
Entrez Gene ID: | 100873949 |
Gene Name: | IPO9 antisense RNA 1 |
Gene Aliases: |
- |
Location: |
Chr.1:201688256-201829559 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| IPO9-AS1 | NR_046696.1 | 1 | 210 |
More Information
Set Membership: |
|

Genomic Map