Assay Details
Target Gene Details
Entrez Gene ID: | 55732 |
Gene Name: | chromosome 1 open reading frame 112 |
Gene Aliases: |
- |
Location: |
Chr.1:169794730-169854080 on Build GRCh38 |
Assay Gene Location: | Within Exon 29 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| C1orf112 | XM_005245317.4 | 24 | 3380 | XP_005245374.1 |
| XM_011509735.2 | 21 | 2990 | XP_011508037.1 | |
| XM_017001724.1 | 23 | 3252 | XP_016857213.1 | |
| XM_017001725.1 | 22 | 3113 | XP_016857214.1 | |
| AK023532.1 | 14 | 2243 | BAB14600.1 |
Target Gene Details
Entrez Gene ID: | 57147 |
Gene Name: | SCY1 like pseudokinase 3 |
Gene Aliases: |
PACE-1, PACE1 |
Location: |
Chr.1:169853074-169894267 on Build GRCh38 |
Assay Gene Location: | Within Exon 16 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SCYL3 | NM_020423.6 | 13 | 2469 | NP_065156.5 |
| NM_181093.3 | 14 | 2631 | NP_851607.2 | |
| XM_006711465.1 | 14 | 2580 | XP_006711528.1 | |
| XM_011509801.1 | 14 | 2720 | XP_011508103.1 | |
| XM_011509802.1 | 14 | 2580 | XP_011508104.1 | |
| XM_011509803.1 | 13 | 2483 | XP_011508105.1 | |
| XM_017001862.1 | 13 | 2418 | XP_016857351.1 | |
| XM_017001863.1 | 13 | 2558 | XP_016857352.1 | |
| XM_017001864.1 | 13 | 2418 | XP_016857353.1 | |
| XM_017001865.1 | 13 | 2517 | XP_016857354.1 | |
| AF540957.1 | 12 | 2299 | AAN23123.1 | |
| AK292507.1 | 14 | 2577 | ||
| AL117233.1 | 13 | 2352 | CAB55300.1 | |
| AY144493.1 | 13 | 2461 | AAN41656.1 | |
| BC014662.1 | 13 | 2406 | AAH14662.1 | |
| BX647352.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2758976 | Chr.1:169812865 - 169916045 on Build GRCh38 | Gain+Loss |
|
More Information
Set Membership: |
|

Genomic Map