Assay Details
Target Gene Details
Entrez Gene ID: | 1136 |
Gene Name: | cholinergic receptor nicotinic alpha 3 subunit |
Gene Aliases: |
LNCR2, NACHRA3, PAOD2 |
Location: |
Chr.15:78593052-78621295 on Build GRCh38 |
Assay Gene Location: | Within Exon 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CHRNA3 | NM_001166694.1 | 6 | 1951 | NP_001160166.1 |
| NR_046313.1 | 8 | 2242 | ||
| AJ584707.1 | 3 | 352 | CAE48369.1 | |
| AJ584708.1 | 2 | 254 | CAE48370.1 | |
| BC006114.1 | 6 | 1951 | AAH06114.1 | |
| BT006646.1 | AAP35292.1 | |||
| M37981.1 | 8 | 1804 | AAA59942.1 |
Target Gene Details
Entrez Gene ID: | 1138 |
Gene Name: | cholinergic receptor nicotinic alpha 5 subunit |
Gene Aliases: |
LNCR2 |
Location: |
Chr.15:78565520-78595269 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 5 - Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CHRNA5 | NM_000745.3 | NP_000736.2 | ||
| NM_001307945.1 | NP_001294874.1 | |||
| XM_005254142.3 | XP_005254199.1 | |||
| XM_017021881.1 | XP_016877370.1 | |||
| AF385586.1 | ||||
| AJ584705.1 | CAE48367.1 | |||
| AJ584706.1 | CAE48368.1 | |||
| BC033639.1 | AAH33639.1 | |||
| BX332469.2 | ||||
| M83712.1 | AAA58357.1 | |||
| U62434.1 | AAB40112.1 | |||
| Y08419.1 | CAA69696.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv518840 | Chr.15:78578946 - 78594829 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map