Assay Details
Target Gene Details
Entrez Gene ID: | 116535 |
Gene Name: | MAS related GPR family member F |
Gene Aliases: |
GPR140, GPR168, MRGF, RTA |
Location: |
Chr.11:69004375-69013382 on Build GRCh38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MRGPRF | NM_001098515.1 | 3 | 1813 | NP_001091985.1 |
| NM_145015.4 | 3 | 1802 | NP_659452.3 | |
| XM_017017170.1 | 3 | 2311 | XP_016872659.1 | |
| AA936120.1 | ||||
| AK075450.1 | 3 | 1689 | BAC11628.1 | |
| AK075492.1 | 4 | 1153 | BAC11649.1 | |
| AK098317.1 | 3 | 1623 | BAC05284.1 | |
| AK122669.1 | 3 | 1687 | ||
| AK131068.1 | 2 | 1405 | ||
| AL049451.1 | 1 | 341 | ||
| BC016964.1 | 3 | 1620 | AAH16964.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv555299 | Chr.11:69002565 - 69053408 on Build GRCh38 | Loss |
|
| nsv832196 | Chr.11:68995114 - 69165820 on Build GRCh38 | Loss |
|
| nsv527120 | Chr.11:68990445 - 69013602 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map