Assay Details
Target Gene Details
Entrez Gene ID: | 57824 |
Gene Name: | histocompatibility minor HB-1 |
Gene Aliases: |
HB-1, HB-1Y, HLA-HB1 |
Location: |
Chr.5:143812161-143820719 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| HMHB1 | NM_021182.1 | 1 | 81 | NP_067005.1 |
| AF103884.1 | 1 | 81 | AAC78642.1 | |
| BC104412.1 | ||||
| BC104413.1 |
More Information
Set Membership: |
|

Genomic Map