Assay Details
Target Gene Details
Entrez Gene ID: | 113612 |
Gene Name: | cytochrome P450 family 2 subfamily U member 1 |
Gene Aliases: |
P450TEC, SPG49, SPG56 |
Location: |
Chr.4:107931549-107953461 on Build GRCh38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CYP2U1 | NM_183075.2 | 2 | 922 | NP_898898.1 |
| XM_005262717.2 | 2 | 988 | XP_005262774.1 | |
| XM_005262720.2 | XP_005262777.1 | |||
| AK290002.1 | 2 | 927 | ||
| AK303284.1 | 3 | 727 | ||
| AK304724.1 | 3 | 800 | ||
| AY343323.1 | 2 | 878 | AAQ21380.1 | |
| BC132767.1 | 2 | 887 | ||
| BC136483.1 | 2 | 887 | ||
| BX354124.2 | 2 | 859 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv461611 | Chr.4:107897601 - 107966289 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map