Assay Details
Target Gene Details
Entrez Gene ID: | 6629 |
Gene Name: | small nuclear ribonucleoprotein polypeptide B2 |
Gene Aliases: |
Msl1, U2B'' |
Location: |
Chr.20:16729964-16741772 on Build GRCh38 |
Assay Gene Location: | Within Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SNRPB2 | NM_003092.4 | 7 | 983 | NP_003083.1 |
| NM_198220.2 | 7 | 883 | NP_937863.1 | |
| AA953215.1 | 1 | 319 | ||
| AI741327.1 | 1 | 710 | ||
| BC018022.1 | 7 | 935 | AAH18022.1 | |
| BC036737.2 | 7 | 842 | AAH36737.1 | |
| BM772256.1 | 2 | 276 | ||
| M15841.1 | 7 | 872 | AAA36796.1 |
More Information
Set Membership: |
|

Genomic Map