Assay Details
Target Gene Details
Entrez Gene ID: | 10525 |
Gene Name: | hypoxia up-regulated 1 |
Gene Aliases: |
GRP-170, Grp170, HSP12A, ORP-150, ORP150 |
Location: |
Chr.11:119044185-119057246 on Build GRCh38 |
Assay Gene Location: | Within Exon 27 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| HYOU1 | NM_001130991.2 | 26 | 3515 | NP_001124463.1 |
| NM_006389.4 | 26 | 3648 | NP_006380.1 | |
| XM_005271392.3 | 26 | 3512 | XP_005271449.1 | |
| XM_005271393.3 | 26 | 3492 | XP_005271450.1 | |
| XM_005271394.3 | 26 | 3559 | XP_005271451.1 | |
| XM_017017095.1 | 26 | 3547 | XP_016872584.1 | |
| XM_017017096.1 | 26 | 3565 | XP_016872585.1 | |
| XM_017017097.1 | 26 | 3488 | XP_016872586.1 | |
| AK127324.1 | 25 | 3520 | ||
| AK297260.1 | 9 | 1041 | ||
| AK299648.1 | 19 | 2442 | ||
| BC004560.2 | 7 | 581 | AAH04560.1 | |
| BC072436.1 | AAH72436.1 | |||
| BE791468.1 | 1 | 266 | ||
| CR749610.1 | 4 | 585 | CAH18405.1 | |
| U65785.1 | 26 | 3516 | AAC50947.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv952745 | Chr.11:119041291 - 119047288 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map