Assay Details
Target Gene Details
Entrez Gene ID: | 5093 |
Gene Name: | poly(rC) binding protein 1 |
Gene Aliases: |
HEL-S-85, HNRPE1, HNRPX, hnRNP-E1, hnRNP-X |
Location: |
Chr.2:70087453-70089203 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PCBP1 | NM_006196.3 | 1 | 1130 | NP_006187.2 |
| AK130439.1 | 1 | 811 | ||
| AK314225.1 | 1 | 997 | ||
| BC039742.1 | 1 | 991 | AAH39742.1 | |
| FJ224329.1 | 1 | 991 | ||
| U24223.1 | 1 | 1016 | AAA91317.1 | |
| X78137.1 | 1 | 869 | CAA55016.1 | |
| Z29505.1 | 1 | 924 | CAA82631.1 |
More Information
Set Membership: |
|

Genomic Map