Assay Details
Target Gene Details
Entrez Gene ID: | 27075 |
Gene Name: | tetraspanin 13 |
Gene Aliases: |
NET-6, NET6, TM4SF13 |
Location: |
Chr.7:16753726-16784536 on Build GRCh38 |
Assay Gene Location: | Within Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TSPAN13 | NM_014399.3 | 6 | 930 | NP_055214.1 |
| AF100759.1 | 6 | 850 | AAD43023.1 | |
| AF120265.1 | 6 | 756 | AAD17294.1 | |
| AK026587.1 | 6 | 894 | ||
| AK075475.1 | 6 | 874 | ||
| AK093487.1 | 6 | 891 | ||
| AY358995.1 | 6 | 861 | AAQ89354.1 | |
| BC033863.1 | 6 | 782 | AAH33863.1 | |
| BX647338.1 | 1 | 1053 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| dgv6289n100 | Chr.7:16230707 - 17452994 on Build GRCh38 | Gain |
|
| nsv1028125 | Chr.7:16489535 - 16807601 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map