Assay Details
Target Gene Details
Entrez Gene ID: | 10634 |
Gene Name: | growth arrest specific 2 like 1 |
Gene Aliases: |
GAR22 |
Location: |
Chr.22:29306618-29312789 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 3 - Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| GAS2L1 | NM_001278730.1 | NP_001265659.1 | ||
| NM_006478.4 | NP_006469.2 | |||
| NM_152236.2 | NP_689422.1 | |||
| XM_011529824.1 | XP_011528126.1 | |||
| XM_011529825.1 | XP_011528127.1 | |||
| XM_017028533.1 | XP_016884022.1 | |||
| AK093386.1 | BAC04150.1 | |||
| AK122829.1 | ||||
| AK223602.1 | BAD97322.1 | |||
| AK225484.1 | ||||
| AK301715.1 | ||||
| BC001782.1 | AAH01782.1 | |||
| BC007624.1 | AAH07624.1 | |||
| BC011047.1 | AAH11047.1 | |||
| Y07846.1 | CAA69174.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv834171 | Chr.22:29230413 - 29433855 on Build GRCh38 | Loss |
|
| nsv829154 | Chr.22:29221496 - 29328012 on Build GRCh38 | Gain |
|
| nsv829155 | Chr.22:29268407 - 29328012 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map