Assay Details
Target Gene Details
Entrez Gene ID: | 7070 |
Gene Name: | Thy-1 cell surface antigen |
Gene Aliases: |
CD90, CDw90 |
Location: |
Chr.11:119417941-119424985 on Build GRCh38 |
Assay Gene Location: | Within Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| THY1 | NM_001311160.1 | 4 | 2681 | NP_001298089.1 |
| NM_001311162.1 | 4 | 1817 | NP_001298091.1 | |
| NM_006288.4 | 4 | 2745 | NP_006279.2 | |
| AF261093.1 | AAG13904.1 | |||
| AK057865.1 | ||||
| AK090996.1 | ||||
| AK094374.1 | ||||
| AL161958.1 | CAB82306.1 | |||
| BE672723.1 | 1 | 284 | ||
| BM504140.1 | 1 | 261 |
Target Gene Details
Entrez Gene ID: | 100499227 |
Gene Name: | USP2 antisense RNA 1 (head to head) |
Gene Aliases: |
THY1-AS1 |
Location: |
Chr.11:119381778-119499233 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| USP2-AS1 | NR_034160.1 | |||
| AK097820.1 | ||||
| AL559224.3 | ||||
| BU171165.1 | ||||
| BU176864.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv832284 | Chr.11:119366940 - 119537915 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map