Assay Details
Target Gene Details
Entrez Gene ID: | 160760 |
Gene Name: | PTC7 protein phosphatase homolog |
Gene Aliases: |
TA-PP2C, TAPP2C |
Location: |
Chr.12:110534432-110583259 on Build GRCh38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PPTC7 | NM_139283.1 | 3 | 759 | NP_644812.1 |
| AF385435.1 | 3 | 759 | AAM43836.1 | |
| AK124744.1 | 4 | 603 | ||
| AY357944.1 | 3 | 528 | AAQ57274.1 | |
| BC111551.1 | 4 | 561 | AAI11552.1 |
More Information
Set Membership: |
|

Genomic Map