Assay Details
Target Gene Details
Entrez Gene ID: | 51065 |
Gene Name: | ribosomal protein S27 like |
Gene Aliases: |
- |
Location: |
Chr.15:63153340-63157542 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| RPS27L | NM_015920.3 | NP_057004.1 | ||
| AF070668.1 | AAD20974.1 | |||
| AK024591.1 | ||||
| AW013916.1 | ||||
| BC003667.1 | AAH03667.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3636712 | Chr.15:63088307 - 63255422 on Build GRCh38 | Gain |
|
| esv27033 | Chr.15:63087706 - 63253263 on Build GRCh38 | Gain+Loss |
|
| nsv428305 | Chr.15:63040203 - 63322630 on Build GRCh38 | Gain |
|
| nsv1035441 | Chr.15:62406037 - 63349997 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map