Assay Details
Target Gene Details
Entrez Gene ID: | 26658 |
Gene Name: | olfactory receptor family 7 subfamily C member 2 |
Gene Aliases: |
CIT-HSP-87M17, OR19-18, OR7C3 |
Location: |
Chr.19:14941489-14942448 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| OR7C2 | NM_012377.1 | 1 | 67 | NP_036509.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv961190 | Chr.19:14941526 - 14942220 on Build GRCh38 | Duplication |
|
| nsv1063744 | Chr.19:14906349 - 14999775 on Build GRCh38 | Gain |
|
| nsv510756 | Chr.19:14926456 - 14952143 on Build GRCh38 | Deletion |
|
| nsv2424 | Chr.19:14934808 - 14978815 on Build GRCh38 | Deletion |
|
| esv3893167 | Chr.19:14610447 - 14950404 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map