Assay Details
Target Gene Details
Entrez Gene ID: | 10595 |
Gene Name: | endoplasmic reticulum to nucleus signaling 2 |
Gene Aliases: |
IRE1-BETA, IRE1b, IRE2p, hIRE2p |
Location: |
Chr.16:23690304-23713500 on Build GRCh38 |
Assay Gene Location: | Within Exon 24 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ERN2 | NM_001308220.1 | 21 | 2960 | NP_001295149.2 |
| NM_033266.3 | 22 | 3116 | NP_150296.4 | |
| XM_011545708.1 | 23 | 2942 | XP_011544010.1 | |
| XM_011545709.1 | 24 | 2993 | XP_011544011.1 | |
| AB047079.1 | BAB21297.1 | |||
| AK131280.1 | 22 | 3116 | BAD18455.1 | |
| BU608100.1 | 1 | 519 | ||
| EF560720.1 | 21 | 2687 | ||
| EF560739.1 | 22 | 2847 |
More Information
Set Membership: |
|

Genomic Map