Assay Details
Target Gene Details
Entrez Gene ID: | 51727 |
Gene Name: | cytidine/uridine monophosphate kinase 1 |
Gene Aliases: |
CK, CMK, CMPK, UMK, UMP-CMPK, UMPK |
Location: |
Chr.1:47333797-47378839 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 3 - Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CMPK1 | NM_001136140.1 | NP_001129612.1 | ||
| NM_016308.2 | NP_057392.1 | |||
| NR_046394.1 | ||||
| AF070416.1 | AAF17709.1 | |||
| AF087865.1 | AAP97174.1 | |||
| AF110643.1 | AAD48583.1 | |||
| AF112216.1 | AAF17204.1 | |||
| AF259961.1 | AAG22609.1 | |||
| AK025258.1 | ||||
| AK223014.1 | BAD96734.1 | |||
| AK293238.1 | ||||
| AK294294.1 | ||||
| AK298502.1 | ||||
| AK312693.1 | ||||
| BC014961.1 | AAH14961.1 | |||
| DB494965.1 |
More Information
Set Membership: |
|

Genomic Map