Assay Details
Target Gene Details
Entrez Gene ID: | 890 |
Gene Name: | cyclin A2 |
Gene Aliases: |
CCN1, CCNA |
Location: |
Chr.4:121816444-121823933 on Build GRCh38 |
Assay Gene Location: | Within Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CCNA2 | NM_001237.3 | 8 | 2457 | NP_001228.1 |
Target Gene Details
Entrez Gene ID: | 5393 |
Gene Name: | exosome component 9 |
Gene Aliases: |
PM/Scl-75, PMSCL1, RRP45, Rrp45p, p5, p6 |
Location: |
Chr.4:121801295-121817021 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 13 - Exon 14 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| EXOSC9 | NM_001034194.1 | NP_001029366.1 | ||
| NM_005033.2 | NP_005024.2 | |||
| AJ505989.1 | CAD44530.1 | |||
| AJ517294.1 | CAD56889.1 | |||
| AK307548.1 | ||||
| BC142978.1 | ||||
| BU615347.1 | ||||
| BX649106.1 | ||||
| M58460.1 | AAA58384.1 | |||
| U09215.1 | AAA18832.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv997539 | Chr.4:121622949 - 122353111 on Build GRCh38 | Gain |
|
| esv3602016 | Chr.4:121811764 - 121818497 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map