Assay Details
Target Gene Details
Entrez Gene ID: | 407004 |
Gene Name: | microRNA 22 |
Gene Aliases: |
MIRN22, hsa-mir-22, miR-22 |
Location: |
Chr.17:1713903-1713987 on Build GRCh38 |
Assay Gene Location: | Overlaps - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MIR22 | NR_029494.1 |
Target Gene Details
Entrez Gene ID: | 84981 |
Gene Name: | MIR22 host gene |
Gene Aliases: |
C17orf91 |
Location: |
Chr.17:1711504-1716272 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 2 - Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MIR22HG | NR_028502.1 | |||
| NR_028503.1 | ||||
| NR_028504.1 | ||||
| NR_028505.1 | ||||
| BC119720.1 | ||||
| BC119721.1 | ||||
| BI756450.1 | ||||
| BM755383.1 | ||||
| BX509423.1 | ||||
| BX640650.1 | ||||
| BX648321.1 | ||||
| DA543295.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv9490 | Chr.17:1653428 - 1828993 on Build GRCh38 | Loss |
|
| nsv470568 | Chr.17:1708905 - 1748486 on Build GRCh38 | Loss |
|
| nsv833335 | Chr.17:1713955 - 1827844 on Build GRCh38 | Gain+Loss |
|
More Information
Additional Information:
Set Membership: |
|

Genomic Map