Assay Details
Target Gene Details
Entrez Gene ID: | 91608 |
Gene Name: | RAS like family 10 member B |
Gene Aliases: |
RRP17, VTS58635 |
Location: |
Chr.17:35731660-35743521 on Build GRCh38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| RASL10B | NM_033315.3 | 4 | 2308 | NP_201572.1 |
| XM_017025300.1 | 5 | 3202 | XP_016880789.1 | |
| XM_017025301.1 | 3 | 2942 | XP_016880790.1 | |
| AK122652.1 | 4 | 2308 | ||
| BC041133.1 | 4 | 2229 | AAH41133.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3892999 | Chr.17:35252863 - 35925533 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map