Assay Details
Target Gene Details
Entrez Gene ID: | 1524 |
Gene Name: | C-X3-C motif chemokine receptor 1 |
Gene Aliases: |
CCRL1, CMKBRL1, CMKDR1, GPR13, GPRV28, V28 |
Location: |
Chr.3:39263494-39281735 on Build GRCh38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CX3CR1 | NM_001171171.1 | 2 | 1701 | NP_001164642.1 |
| NM_001171172.1 | 2 | 1661 | NP_001164643.1 | |
| NM_001171174.1 | 2 | 1597 | NP_001164645.1 | |
| NM_001337.3 | 2 | 1553 | NP_001328.1 | |
| AK225764.1 | 2 | 1553 | ||
| AK307911.1 | ||||
| BC028078.1 | 2 | 1537 | AAH28078.1 | |
| U20350.1 | 2 | 1548 | AAA91783.1 | |
| U28934.1 | AAA87032.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv3772 | Chr.3:39240511 - 39273967 on Build GRCh38 | Insertion |
|
More Information
Set Membership: |
|

Genomic Map