Assay Details
Target Gene Details
Entrez Gene ID: | 140 |
Gene Name: | adenosine A3 receptor |
Gene Aliases: |
A3AR |
Location: |
Chr.1:111499429-111504121 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ADORA3 | NM_000677.3 | 1 | 306 | NP_000668.1 |
| NM_001302678.1 | 1 | 306 | NP_001289607.1 | |
| NM_001302679.1 | 1 | 306 | NP_001289608.1 |
Target Gene Details
Entrez Gene ID: | 57413 |
Gene Name: | transmembrane and immunoglobulin domain containing 3 |
Gene Aliases: |
AD026 |
Location: |
Chr.1:111483348-111564003 on Build GRCh38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TMIGD3 | NM_001081976.2 | NP_001075445.1 | ||
| NM_001302680.1 | NP_001289609.1 | |||
| NM_020683.6 | 1 | 306 | NP_065734.5 | |
| BC041707.1 | AAH41707.1 | |||
| BC064411.1 | AAH64411.1 | |||
| BI828255.1 | ||||
| HY011888.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv463250 | Chr.1:111195294 - 111519590 on Build GRCh38 | Loss |
|
| nsv2410 | Chr.1:111486928 - 111510250 on Build GRCh38 | Insertion |
|
More Information
Set Membership: |
|

Genomic Map