Assay Details
Target Gene Details
Entrez Gene ID: | 5471 |
Gene Name: | phosphoribosyl pyrophosphate amidotransferase |
Gene Aliases: |
ATASE, GPAT, PRAT |
Location: |
Chr.4:56393362-56435636 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 8 - Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PPAT | NM_002703.4 | NP_002694.3 | ||
| XM_011534387.1 | XP_011532689.1 | |||
| AB209246.1 | 1 | 112 | BAD92483.1 | |
| AK222759.1 | BAD96479.1 | |||
| AK290942.1 | ||||
| AK312297.1 | ||||
| BC004200.2 | AAH04200.1 | |||
| D13757.1 | BAA02903.1 | |||
| U00238.1 | AAC27345.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1161086 | Chr.4:56310907 - 56510457 on Build GRCh38 | Duplication |
|
| nsv4347 | Chr.4:56395238 - 56414429 on Build GRCh38 | Deletion |
|
| dgv5213n100 | Chr.4:56076751 - 56538345 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map