Assay Details
Target Gene Details
Entrez Gene ID: | 8689 |
Gene Name: | keratin 36 |
Gene Aliases: |
HA6, KRTHA6, hHa6 |
Location: |
Chr.17:41486136-41489864 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 4 - Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| KRT36 | NM_003771.4 | NP_003762.1 | ||
| BC043581.1 | AAH43581.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv510711 | Chr.17:41455775 - 41574964 on Build GRCh38 | Deletion |
|
| nsv524606 | Chr.17:41483526 - 41510277 on Build GRCh38 | Loss |
|
| nsv483014 | Chr.17:41339595 - 41514244 on Build GRCh38 | Loss |
|
| nsv526466 | Chr.17:41480992 - 41534847 on Build GRCh38 | Gain |
|
| esv2758690 | Chr.17:41145123 - 41740591 on Build GRCh38 | Gain+Loss |
|
More Information
Set Membership: |
|

Genomic Map