Assay Details
Target Gene Details
Entrez Gene ID: | 57182 |
Gene Name: | ankyrin repeat domain 50 |
Gene Aliases: |
- |
Location: |
Chr.4:124664048-124712732 on Build GRCh38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ANKRD50 | NM_001167882.1 | 3 | 2149 | NP_001161354.1 |
| NM_020337.2 | 4 | 3424 | NP_065070.1 | |
| XM_017008471.1 | 3 | 4157 | XP_016863960.1 | |
| AB033049.2 | 1 | 1368 | BAA86537.2 | |
| AK091516.1 | 1 | 1440 | ||
| AK127496.1 | 3 | 2275 | BAC87007.1 | |
| AK291068.1 | 4 | 3424 | ||
| AK295135.1 | 3 | 2149 | ||
| BC024725.1 | 1 | 448 | AAH24725.1 | |
| BX640674.1 | 3 | 1941 | CAE45806.1 | |
| KU178729.1 | 1 | 328 | ||
| KU178730.1 | 1 | 328 |
More Information
Set Membership: |
|

Genomic Map