Assay Details
Target Gene Details
Entrez Gene ID: | 5911 |
Gene Name: | RAP2A, member of RAS oncogene family |
Gene Aliases: |
K-REV, KREV, RAP2, RbBP-30 |
Location: |
Chr.13:97434221-97467998 on Build GRCh38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| RAP2A | NM_021033.6 | 2 | 1932 | NP_066361.1 |
| AF205602.1 | AAN71845.1 | |||
| AK094586.1 | 1 | 663 | ||
| BC041333.2 | 2 | 1933 | AAH41333.1 | |
| BC070031.1 | 2 | 1690 | AAH70031.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv832694 | Chr.13:97278307 - 97485628 on Build GRCh38 | Gain |
|
More Information
Additional Information:
For this assay, SNP(s) [rs113677896] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map