Assay Details
Target Gene Details
Entrez Gene ID: | 1758 |
Gene Name: | dentin matrix acidic phosphoprotein 1 |
Gene Aliases: |
ARHP, ARHR, DMP-1 |
Location: |
Chr.4:87650302-87664361 on Build GRCh38 |
Assay Gene Location: | Within Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| DMP1 | NM_001079911.2 | 5 | 1383 | NP_001073380.1 |
| NM_004407.3 | 6 | 1431 | NP_004398.1 | |
| XM_011531705.2 | 6 | 2306 | XP_011530007.1 | |
| XM_011531706.2 | 5 | 2258 | XP_011530008.1 | |
| BC130581.1 | 5 | 1353 | ||
| BC132865.1 | 5 | 1353 | ||
| U65378.1 | 5 | 1378 | AAB87728.1 | |
| U89012.1 | 6 | 1426 | AAC51332.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3601281 | Chr.4:87609369 - 87722193 on Build GRCh38 | Loss |
|
| nsv1002789 | Chr.4:87318188 - 87685333 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map