Assay Details
Target Gene Details
Entrez Gene ID: | 10011 |
Gene Name: | steroid receptor RNA activator 1 |
Gene Aliases: |
SRA, SRAP, STRAA1, pp7684 |
Location: |
Chr.5:140550067-140558093 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SRA1 | NM_001035235.3 | 1 | 412 | NP_001030312.2 |
| NR_045586.1 | 1 | 412 | ||
| AF293024.1 | AAG02114.1 | |||
| AF293025.1 | AAG02115.1 | |||
| AF293026.1 | AAG02116.1 | |||
| AF318361.1 | 1 | 413 | AAL55868.1 | |
| AK054960.1 | 1 | 629 | ||
| CX165918.1 | ||||
| DQ286291.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv599825 | Chr.5:140556654 - 140626201 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map