Assay Details
Target Gene Details
Entrez Gene ID: | 124989 |
Gene Name: | EF-hand calcium binding domain 13 |
Gene Aliases: |
C17orf57 |
Location: |
Chr.17:47323961-47441312 on Build GRCh38 |
Assay Gene Location: | Within Exon 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| EFCAB13 | NM_001195192.1 | NP_001182121.1 | ||
| NM_152347.4 | 9 | 1031 | NP_689560.3 | |
| AK097661.1 | BAC05135.1 | |||
| BC036407.2 | 9 | 1011 | AAH36407.1 | |
| BC037876.1 | AAH37876.1 | |||
| BX537519.1 | 5 | 4251 | CAD97771.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv516872 | Chr.17:47341773 - 47624914 on Build GRCh38 | Gain+Loss |
|
| dgv3261n100 | Chr.17:47339172 - 47441706 on Build GRCh38 | Loss |
|
| nsv470590 | Chr.17:47341772 - 47469711 on Build GRCh38 | Loss |
|
More Information
Additional Information:
For this assay, SNP(s) [rs71377306] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map