Assay Details
Target Gene Details
Entrez Gene ID: | 407 |
Gene Name: | arrestin 3 |
Gene Aliases: |
ARRX, cArr |
Location: |
Chr.X:70268335-70281840 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 1 - Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ARR3 | NM_004312.2 | NP_004303.2 | ||
| AF033105.1 | AAB84302.1 | |||
| BC012096.1 | AAH12096.1 | |||
| BG472156.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv525258 | Chr.X:70258925 - 70282204 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map