Assay Details
Target Gene Details
Entrez Gene ID: | 51131 |
Gene Name: | PHD finger protein 11 |
Gene Aliases: |
APY, BCAP, IGEL, IGER, IGHER, NY-REN-34, NYREN34 |
Location: |
Chr.13:49495610-49528992 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 3 - Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PHF11 | NM_001040443.2 | NP_001035533.1 | ||
| NM_001040444.2 | NP_001035534.1 | |||
| NR_135322.1 | ||||
| NR_135323.1 | ||||
| AB011031.1 | BAA32101.1 | |||
| AF155105.1 | AAD42871.1 | |||
| AK026228.1 | ||||
| AK225986.1 | ||||
| AK293242.1 | ||||
| AK300410.1 | ||||
| AL552215.3 | ||||
| BC017212.2 | AAH17212.2 | |||
| BX461254.2 | ||||
| DA652444.1 | ||||
| DB050568.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv561613 | Chr.13:49439486 - 49512130 on Build GRCh38 | Loss |
|
| nsv523648 | Chr.13:49431308 - 49512130 on Build GRCh38 | Gain |
|
| dgv3139n54 | Chr.13:49468847 - 49512130 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map