Assay Details
Target Gene Details
Entrez Gene ID: | 140711 |
Gene Name: | TBC/LysM-associated domain containing 2 |
Gene Aliases: |
C20orf118 |
Location: |
Chr.20:36873071-36894235 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 3 - Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TLDC2 | NM_001304783.1 | NP_001291712.1 | ||
| NM_080628.2 | NP_542195.1 | |||
| XM_017027672.1 | XP_016883161.1 | |||
| XM_017027673.1 | XP_016883162.1 | |||
| XM_017027674.1 | XP_016883163.1 | |||
| AK123544.1 | ||||
| BC127688.1 | ||||
| BC130646.1 | ||||
| BC130648.1 | ||||
| BY994247.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3645741 | Chr.20:36801699 - 36878314 on Build GRCh38 | Gain |
|
| nsv1057845 | Chr.20:36855609 - 36916270 on Build GRCh38 | Gain |
|

Genomic Map