Assay Details
Target Gene Details
Entrez Gene ID: | 84532 |
Gene Name: | acyl-CoA synthetase short-chain family member 1 |
Gene Aliases: |
ACAS2L, ACECS1, AceCS2L |
Location: |
Chr.20:25006230-25058182 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 14 - Exon 15 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ACSS1 | NM_001252675.1 | NP_001239604.1 | ||
| NM_001252676.1 | NP_001239605.1 | |||
| NM_001252677.1 | NP_001239606.1 | |||
| NM_032501.3 | NP_115890.2 | |||
| XM_006723659.1 | XP_006723722.1 | |||
| XM_011529388.1 | XP_011527690.1 | |||
| AB058749.1 | BAB47475.1 | |||
| AK027817.1 | BAB55390.1 | |||
| AK090851.1 | BAC03530.1 | |||
| AK092295.1 | BAC03853.1 | |||
| AK125058.1 | BAC86035.1 | |||
| AK125923.1 | ||||
| AK127566.1 | ||||
| AK296306.1 | ||||
| AK314440.1 | ||||
| AL832939.1 | CAH56320.1 | |||
| AL833041.1 | CAH56303.1 | |||
| BC039261.1 | AAH39261.1 | |||
| BC044588.1 | AAH44588.1 | |||
| BC055008.1 | AAH55008.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv585745 | Chr.20:24951392 - 25026453 on Build GRCh38 | Loss |
|
| nsv1056345 | Chr.20:24838934 - 25187523 on Build GRCh38 | Gain |
|
| esv3645552 | Chr.20:24494271 - 25093181 on Build GRCh38 | Gain |
|
| nsv522873 | Chr.20:23923336 - 26134874 on Build GRCh38 | Gain |
|
| esv3645567 | Chr.20:24879101 - 25011663 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map