Assay Details
Target Gene Details
Entrez Gene ID: | 55608 |
Gene Name: | ankyrin repeat domain 10 |
Gene Aliases: |
- |
Location: |
Chr.13:110878540-110915107 on Build GRCh38 |
Assay Gene Location: | Within Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ANKRD10 | NM_017664.3 | 6 | 1364 | NP_060134.2 |
| NR_104587.1 | 7 | 1426 | ||
| AK000100.1 | 6 | 1326 | BAA90945.1 | |
| AK000111.1 | 1 | 657 | BAA90951.1 | |
| AK026017.1 | 2 | 438 | BAB15323.1 | |
| AK094222.1 | ||||
| AK095763.1 | 4 | 1036 | ||
| AK096083.1 | 3 | 734 | ||
| AK123341.1 | 1 | 1199 | ||
| AK125100.1 | 1 | 2334 | ||
| BX641022.1 | 1 | 2362 | CAH56123.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1044340 | Chr.13:110581726 - 111040647 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map