Assay Details
Target Gene Details
Entrez Gene ID: | 7349 |
Gene Name: | urocortin |
Gene Aliases: |
UI, UROC |
Location: |
Chr.2:27307400-27308445 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| UCN | NM_003353.3 | NP_003344.1 | ||
| AI857458.1 | ||||
| BC104470.1 | AAI04471.1 | |||
| BC104471.2 | AAI04472.1 | |||
| BU619919.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv833614 | Chr.2:27263559 - 27452213 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map