Assay Details
Target Gene Details
Entrez Gene ID: | 55609 |
Gene Name: | zinc finger protein 280C |
Gene Aliases: |
SUHW3, ZNF633 |
Location: |
Chr.X:130202699-130268948 on Build GRCh38 |
Assay Gene Location: | Within Exon 14 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ZNF280C | NM_017666.4 | 14 | 1995 | NP_060136.1 |
| XM_006724765.3 | 13 | 1799 | XP_006724828.1 | |
| AK000102.1 | 14 | 1918 | BAA90947.1 | |
| AK290373.1 | 14 | 1946 | ||
| AL133204.1 | 13 | 1783 | CAB61579.1 | |
| AL834333.1 | 14 | 1926 | CAD39001.1 | |
| BC051728.1 | 14 | 1946 | AAH51728.1 | |
| DA441065.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv528290 | Chr.X:129635682 - 130429852 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map