Assay Details
Target Gene Details
Entrez Gene ID: | 9338 |
Gene Name: | transcription elongation factor A like 1 |
Gene Aliases: |
SIIR, WEX9, p21, pp21 |
Location: |
Chr.X:103628720-103630953 on Build GRCh38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TCEAL1 | NM_001006639.1 | 3 | 730 | NP_001006640.1 |
| NM_001006640.1 | 3 | 742 | NP_001006641.1 | |
| NM_004780.2 | 3 | 742 | NP_004771.2 | |
| BC000809.1 | 3 | 695 | AAH00809.1 | |
| BU602495.1 | ||||
| BX092470.1 | ||||
| CF454849.1 | ||||
| M99701.1 | 3 | 732 | AAA60149.1 |
More Information
Set Membership: |
|

Genomic Map