Assay Details
Target Gene Details
Entrez Gene ID: | 57182 |
Gene Name: | ankyrin repeat domain 50 |
Gene Aliases: |
- |
Location: |
Chr.4:124664048-124712732 on Build GRCh38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ANKRD50 | NM_001167882.1 | 4 | 5829 | NP_001161354.1 |
| NM_020337.2 | 5 | 7104 | NP_065070.1 | |
| XM_017008471.1 | 4 | 7837 | XP_016863960.1 | |
| AB033049.2 | 2 | 5048 | BAA86537.2 | |
| AL110131.1 | 1 | 1188 |
More Information
Set Membership: |
|

Genomic Map