Assay Details
Target Gene Details
Entrez Gene ID: | 1520 |
Gene Name: | cathepsin S |
Gene Aliases: |
- |
Location: |
Chr.1:150730196-150765957 on Build GRCh38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CTSS | NM_001199739.1 | 4 | 644 | NP_001186668.1 |
| NM_004079.4 | 5 | 794 | NP_004070.3 | |
| AK226095.1 | 5 | 656 | ||
| AK301472.1 | 4 | 479 | ||
| AK314482.1 | 5 | 635 | ||
| BC002642.2 | 5 | 595 | AAH02642.1 | |
| CR541676.1 | 4 | 533 | CAG46477.1 | |
| M86553.1 | 4 | 539 | AAA35655.1 | |
| M90696.1 | 5 | 669 | AAC37592.1 | |
| S93414.1 | 4 | 539 | AAB22005.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1008381 | Chr.1:150748778 - 150773082 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map