Assay Details
Target Gene Details
Entrez Gene ID: | 4338 |
Gene Name: | molybdenum cofactor synthesis 2 |
Gene Aliases: |
MCBPE, MOCO1, MOCODB, MPTS |
Location: |
Chr.5:53095679-53109772 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MOCS2 | NM_004531.4 | 1 | 358 | NP_004522.1 |
| NM_176806.3 | NP_789776.1 | |||
| AA252410.1 | 1 | 81 | ||
| AF091871.1 | AAD14598.1 | |||
| AF117815.1 | AAD13296.1 | |||
| AF155659.1 | 1 | 81 | AAF67478.1 | |
| AK312887.1 | ||||
| BC046097.1 | 1 | 44 | AAH46097.1 | |
| CD514049.1 | ||||
| DA967651.1 | 1 | 341 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv830296 | Chr.5:52913698 - 53114423 on Build GRCh38 | Gain |
|
| esv3570141 | Chr.5:53108676 - 53114036 on Build GRCh38 | Loss |
|
| nsv442948 | Chr.5:53108689 - 53113609 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map